missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

eBioscience™ Human sCD44std Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human sCD44std.

$665.55 - $4,334.05

Specifications

For Use With (Application) ELISA
Sample Size 20μL (1:60 prediluted)
Synonym Phagocytic glycoprotein I, Hermes antigen, Epican
Target CD44std, sCD44std
Conjugate HRP
View More Specs

Disclaimers:

For Research Use Only


Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD44std
HRP-Conjugate anti-human sCD44std monoclonal antibody
Human sCD44std Standard, 8ng/mL
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501125297 Life Technologies
LSBMS2092CE
96 tests Each for $665.55
501125298 Life Technologies
LSBMS2092TENCE
10 x 96 tests Each for $4,334.05
Specifications

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Description & Specifications

Specifications

ELISA
Phagocytic glycoprotein I, Hermes antigen, Epican
HRP
0.02ng/mL
20μL (1:60 prediluted)
CD44std, sCD44std
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin), amniotic fluid, urine
Human