Please login to your online account to display your discounted pricing

eBioscience™ Human sCD40L Platinum ELISA Kit, extra sensitive

For the quantitative detection of human sCD40L.

$520.84 - $2,932.34

Specifications

For Use With (Application) ELISA
Sample Size 20 to 50μL
Synonym TRAP, HIGM 1, T cell antigen Gp39, CD154
Target sCD154 (sCD40 Ligand), sCD40
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD40L
HRP-Conjugate anti-human sCD40L monoclonal antibody
Human sCD40L Standard lyophilized, 20ng/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018246

 
life technologies
LSBMS293
96 tests Each for $520.84

5018248

 
life technologies
LSBMS293TEN
10 x 96 tests Each for $2,932.34
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 20 to 50μL
Synonym TRAP, HIGM 1, T cell antigen Gp39, CD154
Target sCD154 (sCD40 Ligand), sCD40
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Sensitivity 0.01ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.