Please login to your online account to display your discounted pricing

eBioscience™ Human sCD26 Platinum ELISA Kit

For the quantitative detection of human sCD26.

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 20μL
Synonym Dipeptidyl peptidase IV, ADCP 2, ADABP, TP 103
Target CD26, sCD26
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD26
Biotin-Conjugate anti-human sCD26 monoclonal antibody
Streptavidin-HRP
Human sCD26 Standard, 500ng/mL upon dilution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018115

 
life technologies
LSBMS235
96 tests Each for $651.05

5018117

 
life technologies
LSBMS235TEN
10 x 96 tests Each for $4,189.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 20μL
Synonym Dipeptidyl peptidase IV, ADCP 2, ADABP, TP 103
Target CD26, sCD26
Conjugate Biotin
Sample Type Cell culture supernatant, serum
Sensitivity 7.3ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.