Learn More
Invitrogen™ sCD134/OX40 (Soluble) Human ELISA Kit
For the quantitative detection of human sCD134/OX40.
$752.00 - $5,240.00
Specifications
Accession Number | P43489 |
---|---|
Assay Range | 78-5,000 pg/mL |
Assay Sensitivity | 1.8 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD134/OX40
Biotin-Conjugate anti-human sCD134/OX40 monoclonal antibody
Streptavidin-HRP
Human sCD134/OX40 Standard lyophilized, 10ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
The Human soluble CD134/OX40 ELISA quantitates Hu CD134/OX40 in human serum, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu CD134/OX40. Principle of the method The Human CD134/OX40 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
P43489 | |
1.8 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
ACT35,CD134,IMD16,OX40,TXGP1L | |
8% | |
HRP | |
Serum, 25 μL; Supernatant, 25 μL | |
OX 40L receptor, ACT 35 | |
1 hr. 20 min. |
78-5,000 pg/mL | |
Biotin | |
Serum, Supernatant | |
ELISA | |
7293 | |
12% | |
Pre-coated 96 well plate, Standard, Standard, Sample Diluent, Biotinylated Detection Antibody, SAV-HRP, SAV-HRP, Lysis Buffer, Lysis Buffer, Stop Solution, Stop Solution | |
RUO | |
2°C to 8°C | |
Human | |
3 hr. 10 min. |
For Research Use Only