Please login to your online account to display your discounted pricing

eBiosciencetrade; Human sAPO-1/Fas Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human sAPO-1/Fas.

$665.55 - $4,334.05

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym CD95, Fas receptor, APT 1, ALPS 1 A, TNFRSF6
Target CD95 (APO-1/Fas), sCD95 (sAPO-1/Fas)
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sAPO-1/Fas
Biotin-Conjugate anti-human sAPO-1/Fas monoclonal antibody
Streptavidin-HRP
Human sAPO-1/Fas Standard lyophilized, 2ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125409

 
ebioscience
LSBMS245CE
96 tests Each for $665.55

501125410

 
ebioscience
LSBMS245TENCE
10 x 96 tests Each for $4,334.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym CD95, Fas receptor, APT 1, ALPS 1 A, TNFRSF6
Target CD95 (APO-1/Fas), sCD95 (sAPO-1/Fas)
Conjugate Biotin
Sample Type Cell culture supernatant, serum
Sensitivity 13.2pg/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.