missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

eBioscience™ Human Resistin Platinum ELISA Kit

For the quantitative detection of human Resistin. Resistin is a hormone secreted by adipose tissue.

$592.68 - $3,813.48

Specifications

For Use With (Application) ELISA
Sample Size 50μL (1:10 prediluted)
Synonym Cysteine-rich secreted protein FIZZ3, ADSF, RETN1
Target Resistin
Conjugate Biotin
View More Specs

Disclaimers:

For Research Use Only


Includes: Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to human Resistin
Biotin-Conjugate anti-human Resistin polyclonal antibody
Streptavidin-HRP
Human Resistin Standard lyophilized, 4000pg/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5017946 Life Technologies
LSBMS2040
96 tests Each for $592.68
5017947 Life Technologies
LSBMS2040TEN
10 x 96 tests Each for $3,813.48
Specifications

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Description & Specifications

Specifications

ELISA
Cysteine-rich secreted protein FIZZ3, ADSF, RETN1
Biotin
3.1pg/mL
50μL (1:10 prediluted)
Resistin
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Human