missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human Pro-MMP-13 Quantikine ELISA Kit, R&D Systems

a 4.5 hour solid-phase ELISA designed to measure pro-MMP-13 in cell culture supernates.

$606.82 - $2,901.06

Specifications

Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 4322
Sample Volume Cell Culture Supernates (50μL)
Sensitivity 21.3pg/mL
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
DM1300 R & D Systems
RDSDM1300
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $606.82
SMP1300 R & D Systems
RDSSMP1300
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $2,901.06
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL)
Sensitivity: 21.3pg/mL
Specificity: Natural and recombinant human pro-MMP-13
Cross Reactivity: Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
4322
21.3pg/mL
Solid Phase Sandwich ELISA
Cell Culture Supernates (50μL)
Human