missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human Pro-MMP-1 Quantikine ELISA Kit, R&D Systems

$624.40 - $2,985.15

Specifications

Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 4312
Sample Volume Cell Culture Supernates (100μL), Serum (100μL), Heparin Plasma (100μL)
Sensitivity 0.095ng/mL
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
DMP100 R & D Systems
RDSDMP100
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $624.40
SMP100 R & D Systems
RDSSMP100
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $2,985.15
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (100μL), Serum (100μL), Heparin Plasma (100μL)
Sensitivity: 0.095ng/mL
Specificity: Natural and recombinant human Pro-MMP-1
Cross Reactivity: Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
4312
0.095ng/mL
Solid Phase Sandwich ELISA
Cell Culture Supernates (100μL), Serum (100μL), Heparin Plasma (100μL)
Human