Please login to your online account to display your discounted pricing

Human PlGF Quantikine ELISA Kit, R&D Systems

$662.25 - $3,178.57

Specifications

Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 5228
Sample Volume Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL), Heparin Plasma (100μL), Citrate Plasma (100μL), Urine (100μL)
Sensitivity 7pg/mL
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity    

DPG00

 
r & d systems
RDSDPG00
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $662.25

SPG00

 
r & d systems
RDSSPG00
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $3,178.57
Description & Specifications

Specifications

Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 5228
Sample Volume Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL), Heparin Plasma (100μL), Citrate Plasma (100μL), Urine (100μL)
Sensitivity 7pg/mL
Species Human

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL), Heparin Plasma (100μL), Citrate Plasma (100μL), Urine (100μL)
Sensitivity: 7pg/mL
Specificity: Natural and recombinant human PlGF
Cross Reactivity: Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.