Please login to your online account to display your discounted pricing

Human Physiology Charts

Explain the major functional systems of human anatomy and physiology.

$820.85 - $833.15

Specifications

Classification Life science
For Use With (Application) Explains major functional systems of Human anatomy, physiology, each main portrait has up to a dozen supplementary figures
Includes Flip Charthead and Tripod Stand
Width (English) 36 in.
View More Specs
Products
Catalog Number Mfr. No. Format Price Quantity    

S65073

 
fisher science education
1400-41-EMD
Charthead with tripod base Each for $820.85

S66212

 
fisher science education
1400-32
Multi-chart spring roller system Each for $833.15
Description & Specifications

Specifications

Classification Life science
For Use With (Application) Explains major functional systems of Human anatomy, physiology, each main portrait has up to a dozen supplementary figures
Includes Flip Charthead and Tripod Stand
Width (English) 36 in.

  • Focus on major body systems by detailing the function and features vital to understanding human biology
  • Each main portrait has up to a dozen supplementary figures from a variety of angles and magnifications Labeled with call-outs pointing directly to key features
  • Color-coded to add emphasis and clarity where appropriate

  • Set of 10 charts includes: The Skeletal System, Muscular System, Brain and Nervous System, Circulatory System, Endocrine System, Respiratory System, Digestive System, Eye and Vision, Ear and Nose, Glands, and Skin and Excretory Organs
  • Set is available on charthead with a tripod base or mounted on a spring roller