missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human Phospho-TOR (S2448) DuoSet IC, R&D Systems

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Manufacturer:  R & D Systems RDSDYC16652

 View more versions of this product

Catalog No. DYC16652


 

Specifications

Description

Kit, ELISA; R&D Systems; Human Phospho-TOR (S2448) DuoSet IC; Two 96-well plates; 1 Kit; Sufficient materials to run ELISAs on at least two 96-well plates

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Description & Specifications

Specifications

ELISA
2475
2 Plates
Human
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for phosphorylated intracellular proteins
1 Kit