Please login to your online account to display your discounted pricing

Human Phospho-Ret DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC65492

 View more versions of this product

Catalog No. DYC65492

  • / Each
 

Description & Specifications

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Gene ID (Entrez) 5979
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Packaging Quantity 2 Plates
Species Human
Quantity 1 Kit