Please login to your online account to display your discounted pricing

Human Phospho-PRAS40 (T246) DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC68902

 View more versions of this product

Catalog No. DYC68902

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Packaging Quantity 2 Plates
Species Human
Gene ID (Entrez) 84335
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Quantity 1 Kit