Please login to your online account to display your discounted pricing

Human Phospho-PDGF R alpha DuoSet IC, R&D Systems

$419.81 - $1,780.05

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC2114E

 
r & d systems
RDSDYC2114E
15 Plates Each for $1,780.05

DYC21145

 
r & d systems
RDSDYC21145
5 Plates Each for $828.14

DYC21142

 
r & d systems
RDSDYC21142
2 Plates Each for $419.81
Description & Specifications

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins