Please login to your online account to display your discounted pricing

Human Phospho-MKK6 (S207/T211) DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC55862

 View more versions of this product

Catalog No. DYC55862

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 5608
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Packaging Quantity 2 Plates
Quantity 1 Kit
Species Human

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.