Please login to your online account to display your discounted pricing

Human Phospho-Mer DuoSet IC, R&D Systems

$419.81 - $1,780.05

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC2579E

 
r & d systems
RDSDYC2579E
15 Plates Each for $1,780.05

DYC25792

 
r & d systems
RDSDYC25792
2 Plates Each for $419.81

DYC25795

 
r & d systems
RDSDYC25795
5 Plates Each for $828.14
Description & Specifications

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins