Please login to your online account to display your discounted pricing

Human Phospho-M-CSF R DuoSet IC, R&D Systems

Contains the basic components required for the development of sandwhich ELISAs to measure tyrosine-phosphorylated in cell lysates.

$428.87 - $1,818.46

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC3268E

 
r & d systems
RDSDYC3268E
15 Plates Each for $1,818.46

DYC32685

 
r & d systems
RDSDYC32685
5 Plates Each for $846.01

DYC32682

 
r & d systems
RDSDYC32682
2 Plates Each for $428.87
Description & Specifications

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human

  • Optimized capture and detection antibody pairings and recommended concentrations save lengthy development time
  • Assay can be customized to your specific needs
  • Economical alternative to Western blot