Please login to your online account to display your discounted pricing

Human Phospho-IFN-gamma R1 DuoSet IC, R&D Systems

$419.81 - $419.81

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC30582

 
r & d systems
RDSDYC30582
2 Plates Each for $419.81
Description & Specifications

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins