Please login to your online account to display your discounted pricing

Human Phospho-IFN-gamma R1 DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC30582

 View more versions of this product

Catalog No. DYC30582

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3459
Quantity 1 Kit
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human
Packaging Quantity 2 Plates