Please login to your online account to display your discounted pricing

Human Phospho-Histone H2AX (S139) DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC22882

 View more versions of this product

Catalog No. DYC22882

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Gene ID (Entrez) 3014
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Packaging Quantity 2 Plates
Quantity 1 Kit
Species Human