Please login to your online account to display your discounted pricing

Human Phospho-HGF R/c-MET DuoSet IC, R&D Systems

$419.81 - $1,818.33

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC2480E

 
r & d systems
RDSDYC2480E
15 Plates Each for $1,818.33

DYC24802

 
r & d systems
RDSDYC24802
2 Plates Each for $419.81

DYC24805

 
r & d systems
RDSDYC24805
5 Plates Each for $828.14
Description & Specifications

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins