Please login to your online account to display your discounted pricing

Human Phospho-HGF R/c-MET DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC24805

 View more versions of this product

Catalog No. DYC24805

  • / Each
 

Description & Specifications

Specifications

Species Human
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 4233
Packaging Quantity 5 Plates