Please login to your online account to display your discounted pricing

Human Phospho-HGF R/c-MET DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC24802

 View more versions of this product

Catalog No. DYC24802

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 4233
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Packaging Quantity 2 Plates
Quantity 1 Kit
Species Human