Please login to your online account to display your discounted pricing

Human Phospho-GSK-3 alpha (S21) DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC41252

 View more versions of this product

Catalog No. DYC41252

  • / Each
 

Description & Specifications

Specifications

Packaging Quantity 2 Plates
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Quantity 1 Kit
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Species Human
Gene ID (Entrez) 2931