missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human Phospho-gp130 DuoSet IC, R&D Systems

$428.87 - $428.87

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
DYC34072 R & D Systems
RDSDYC34072
2 Plates Each for $428.87
Specifications

Description

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins

Description & Specifications

Specifications

ELISA Development Kit for phosphorylated intracellular proteins
Human