Please login to your online account to display your discounted pricing

Human Phospho-gp130 DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC34072

 View more versions of this product

Catalog No. DYC34072

  • / Each
 

Description & Specifications

Specifications

Species Human
Quantity 1 Kit
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Gene ID (Entrez) 3572
Packaging Quantity 2 Plates