Please login to your online account to display your discounted pricing

Human Phospho-FGF R2 alpha DuoSet IC, R&D Systems

$428.87 - $1,857.57

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC6842

 
r & d systems
RDSDYC6842
2 Plates Each for $428.87

DYC684E

 
r & d systems
RDSDYC684E
15 Plates Each for $1,857.57

DYC6845

 
r & d systems
RDSDYC6845
5 Plates Each for $846.01
Description & Specifications

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins