Please login to your online account to display your discounted pricing

Human Phospho-ErbB3/Her3 DuoSet IC, R&D Systems

$419.81 - $1,818.33

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC17695

 
r & d systems
RDSDYC17695
5 Plates Each for $828.14

DYC1769E

 
r & d systems
RDSDYC1769E
15 Plates Each for $1,818.33

DYC17692

 
r & d systems
RDSDYC17692
2 Plates Each for $419.81
Description & Specifications

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins