Please login to your online account to display your discounted pricing

Human Phospho-ErbB2/Her2 (Y1196) DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC44382

 View more versions of this product

Catalog No. DYC44382

  • / Each
 

Description & Specifications

Specifications

Packaging Quantity 2 Plates
Gene ID (Entrez) 2064
Quantity 1 Kit
Species Human
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.