Please login to your online account to display your discounted pricing

Human Phospho-ER alpha/NR3A1 (S118) DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC59542

 View more versions of this product

Catalog No. DYC59542

  • / Each
 

Description & Specifications

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Quantity 1 Kit
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Species Human
Packaging Quantity 2 Plates
Gene ID (Entrez) 2099