Please login to your online account to display your discounted pricing

Human Phospho-EGF R/ErbB1 DuoSet IC, R&D Systems

$419.81 - $1,818.33

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC1095B2

 
r & d systems
RDSDYC1095B2
2 Plates Each for $419.81

DYC1095B5

 
r & d systems
RDSDYC1095B5
5 Plates Each for $828.14

DYC1095BE

 
r & d systems
RDSDYC1095BE
15 Plates Each for $1,818.33
Description & Specifications

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins