Please login to your online account to display your discounted pricing

Human Phospho-DDR1 DuoSet IC, R&D Systems

$419.81 - $828.14

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC58592

 
r & d systems
RDSDYC58592
2 Plates Each for $419.81

DYC58595

 
r & d systems
RDSDYC58595
5 Plates Each for $828.14
Description & Specifications

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Species Human

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins