Please login to your online account to display your discounted pricing

eBioscience Human PEI-Transferrinfection

Nucleic acid delivery system based on transferrin receptor-mediated endocytosis to carry DNA into cells and the gene transfer efficiency of PEI/DNA complexes

$195.75 - $355.25

Specifications

For Use With (Application) Cell Culture
Species Human
View More Specs
Includes:  Transferrin-Polyethylenimine-conjugate, lyophilized, 1 mg/mL upon reconstitution; iron incorporated
Deferrioxamine, lyophilied, 50 mM upon reconstitution
HBS buffer concentrate (20x)
(400mM Hepes, pH 7.3, 3M NaCl)
Products
Catalog Number Mfr. No. Quantity Price Quantity    

50246647

 
ebioscience
LSBMS1003A
16 transferrinfections Each for $195.75

50246646

 
ebioscience
LSBMS1003
32 transferrinfections Each for $355.25
Description & Specifications

Specifications

For Use With (Application) Cell Culture
Species Human

  • Method results in a 30 to 1000-fold enhanced transfection efficiency depending on the cell line
  • An extremely gentle DNA transfection method which employs physiological uptake mechanisms of the cell
  • Transfection efficacy depends on the cell type, the level of surface transferrin receptor expression
  • Very high transfection rates have been shown for the tested tumor cell lines B16F10 melanoma, Neuro 2A neuroblastoma and a variety of primary human melanoma cell lines
  • In other established cell lines, such as HeLa, CHO, Jurkat, K562, HepG2 and COS the PEI-Transferrinfection works with high efficiences, excellent reproducibility and with the advantage of being an extremely gentle procedure