missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PDC (aa 28-80) Control Fragment Recombinant Protein

Catalog No. RP107146
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP107146 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP107146 Supplier Invitrogen™ Supplier No. RP107146
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66472 (PA5-66472. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PDC encodes a phosphoprotein, which is located in the outer and inner segments of the rod cells in the retina. This protein may participate in the regulation of visual phototransduction or in the integration of photoreceptor metabolism. It modulates the phototransduction cascade by interacting with the beta and gamma subunits of the retinal G-protein transducin. PDC is a potential candidate gene for retinitis pigmentosa and Usher syndrome type II. Alternatively spliced transcript variants encoding different isoforms have been identified.

Specifications

Accession Number P20941
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5132
Name Human PDC (aa 28-80) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 33 kDa phototransducing protein; 33 DPTP; G beta gamma binding protein; MEKA; MEKA protein; Pdc; PHD; PhLOP; PhLP; Phosducin; phosducin-like orphan protein; Phototransducing protein, 33 kDa (phosducin); Protein MEKA; Rod photoreceptor 1; Rpr1; RPR-1
Common Name PDC
Gene Symbol PDC
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DWRKFKLESQDSDSIPPSKKEILRQMSSPQSRNGKDSKERVSRKMSIQEYELI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less