Please login to your online account to display your discounted pricing

eBioscience™ Human PD-L2 Platinum ELISA Kit

For quantitative detection of human PD-L2.

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym PDCD1LG2, B7DC, Btdc, CD273, PD-L2, PDCD1L2, PDL2, bA574F11.2
Target PD-L2
Sample Type Serum, plasma (citrate, heparin, EDTA) and cell culture supernatant
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human PD-L2
Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125372

 
life technologies
LSBMS2215
96 tests Each for $651.05

501125373

 
life technologies
LSBMS2215TEN
10 x 96 tests Each for $4,189.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym PDCD1LG2, B7DC, Btdc, CD273, PD-L2, PDCD1L2, PDL2, bA574F11.2
Target PD-L2
Sample Type Serum, plasma (citrate, heparin, EDTA) and cell culture supernatant
Sensitivity 3.65pg/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.