Invitrogen™ eBioscience™ Human PD-L2 Platinum ELISA Kit
For quantitative detection of human PD-L2.
Manufacturer:  Invitrogen™ LSBMS2215TEN
 View more versions of this product
Catalog No. 501125373
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human PD-L2
Description
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
ELISA
3.65pg/mL
10μL
PD-L2
10 x 96 tests
Serum, plasma (citrate, heparin, EDTA) and cell culture supernatant
Human
PDCD1LG2, B7DC, Btdc, CD273, PD-L2, PDCD1L2, PDL2, bA574F11.2
Documents