Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Human Papilloma Virus Type 16-E7 (HPV 16-E7) Ab-1, Mouse Monoclonal Antibody

Recommended for Immunofluorescence and Flow Cytometry, Thermo Scientific™ Human Papilloma Virus Type 16-E7 (HPV 16-E7) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

$238.04 - $1,005.04

Specifications

Antigen Human Papilloma Virus Type 16-E7 (HPV 16-E7) Ab-1
Clone TVG701Y
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS768P

 
thermo scientific™ lab vision™
MS768P
200μg/mL 1mL Each for $890.80

MS768P0

 
thermo scientific™ lab vision™
MS768P0
200μg/mL 100μL Each for $238.04

MS768P1

 
thermo scientific™ lab vision™
MS768P1
200μg/mL 500μL Each for $667.27

MS768P1ABX

 
thermo scientific™ lab vision™
MS768P1ABX
1mg/mL 100μL Each for $741.63

MS768PABX

 
thermo scientific™ lab vision™
MS768PABX
1mg/mL 200μL Each for $1,005.04
Description & Specifications

Specifications

Antigen Human Papilloma Virus Type 16-E7 (HPV 16-E7) Ab-1
Clone TVG701Y
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Infectious Disease
Type Primary
Immunogen Recombinant HPV-16 E7 protein
Isotype IgG2a kappa

Recommended for Immunofluorescence and Flow Cytometry, Thermo Scientific™ Human Papilloma Virus Type 16-E7 (HPV 16-E7) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

E7 is an early protein of type 16 human papilloma virus (HPV 16-E7). Infection with specific types of HPV has been associated with an increased risk of developing cervical neoplasia. HPV types 6 and 11 have been associated with relatively benign diseases such as genital warts but types 16 and 18 are strongly associated with cervical, vaginal, and vulvar malignancies.

Host Species: Mouse

Clone: TVG701Y

Isotype: IgG2a/κ

Species Reactivity: Type 16 of human Papilloma Virus (HPV-16).

Immunogen: Recombinant HPV-16 E7 protein

Molecular Weight: 15kDa

Positive Control: Cervical tissue infected with HPV-16

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting (Not verified)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO