missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Human Papilloma Virus Type 16-E7 (HPV 16-E7) Ab-1, Mouse Monoclonal Antibody

Recommended for Immunofluorescence and Flow Cytometry, Thermo Scientific™ Human Papilloma Virus Type 16-E7 (HPV 16-E7) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS768P1ABX

 View more versions of this product

Catalog No. MS768P1ABX


 

Specifications

Description

Recommended for Immunofluorescence and Flow Cytometry, Thermo Scientific™ Human Papilloma Virus Type 16-E7 (HPV 16-E7) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

E7 is an early protein of type 16 human papilloma virus (HPV 16-E7). Infection with specific types of HPV has been associated with an increased risk of developing cervical neoplasia. HPV types 6 and 11 have been associated with relatively benign diseases such as genital warts but types 16 and 18 are strongly associated with cervical, vaginal, and vulvar malignancies.

Host Species: Mouse

Clone: TVG701Y

Isotype: IgG2a/κ

Species Reactivity: Type 16 of human Papilloma Virus (HPV-16).

Immunogen: Recombinant HPV-16 E7 protein

Molecular Weight: 15kDa

Positive Control: Cervical tissue infected with HPV-16

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting (Not verified)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Human Papilloma Virus Type 16-E7 (HPV 16-E7) Ab-1
Monoclonal
1mg/mL
Concentrated
Recombinant HPV-16 E7 protein
100μL
Infectious Disease
Monoclonal
Immunofluorescence
TVG701Y
Unlabeled
Murine
IgG2a kappa
RUO
Primary
Human