Please login to your online account to display your discounted pricing

Human/Mouse Total Pin1 DuoSet IC, R&D Systems

$428.87 - $428.87

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Species Human, Mouse
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC22942

 
r & d systems
RDSDYC22942
2 Plates Each for $428.87
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Species Human, Mouse

Assay Type: ELISA Development Kit for intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.