missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human/Mouse Total HIF-1 alpha DuoSet IC, R&D Systems

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

$428.87 - $1,818.46

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Species Human, Mouse
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
DYC1935E R & D Systems
RDSDYC1935E
15 Plates Each for $1,818.46
DYC19352 R & D Systems
RDSDYC19352
2 Plates Each for $428.87
DYC19355 R & D Systems
RDSDYC19355
5 Plates Each for $846.01
Specifications

Description

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs See Product Certificates

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for intracellular proteins
Human, Mouse