Please login to your online account to display your discounted pricing

Human/Mouse/Rat Total TC-PTP DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC19302

 View more versions of this product

Catalog No. DYC19302

  • / Each
 

Description & Specifications

Specifications

Species Human, Mouse, Rat
Gene ID (Entrez) 5771
Quantity 1 Kit
Product Type ELISA Development Kit for intracellular proteins
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Format Two 96-well plates