Please login to your online account to display your discounted pricing

Human/Mouse/Rat Total SOD2/Mn-SOD DuoSet IC, R&D Systems

$428.87 - $846.01

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 6648
Species Human, Mouse, Rat
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC34192

 
r & d systems
RDSDYC34192
2 Plates Each for $428.87

DYC34195

 
r & d systems
RDSDYC34195
5 Plates Each for $846.01
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 6648
Species Human, Mouse, Rat

Assay Type: ELISA Development Kit for intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.