Please login to your online account to display your discounted pricing

Human/Mouse/Rat Total PP2A DuoSet IC, R&D Systems

$419.81 - $828.14

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 5515
Species Human, Mouse, Rat
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC16535

 
r & d systems
RDSDYC16535
5 Plates Each for $828.14

DYC16532

 
r & d systems
RDSDYC16532
2 Plates Each for $419.81
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 5515
Species Human, Mouse, Rat

Assay Type: ELISA Development Kit for intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.