Please login to your online account to display your discounted pricing

Human/Mouse/Rat Total Numb DuoSet IC, R&D Systems

$419.81 - $419.81

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Quantity 1 Kit
Species Human, Mouse, Rat
View More Specs
Products
Catalog Number Mfr. No. Gene ID (Entrez) Format Price Quantity    

DYC43382

 
r & d systems
RDSDYC43382
29419 Two 96-well plates Each for $419.81
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Quantity 1 Kit
Species Human, Mouse, Rat

Assay Type: ELISA Development Kit for intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.