Please login to your online account to display your discounted pricing

Human/Mouse/Rat Total JNK Pan Specific DuoSet IC, R&D Systems

$419.81 - $419.81

Specifications

Applications Development of sandwich ELISA to measure JNK in cell lysates
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 kit
Species Human, mouse, rat
View More Specs
Products
Catalog Number Mfr. No. Format Price Quantity    

DYC12052

 
r & d systems
RDSDYC12052
Two 96-well plates Each for $419.81
Description & Specifications

Specifications

Applications Development of sandwich ELISA to measure JNK in cell lysates
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 kit
Species Human, mouse, rat

Assay Type: ELISA Development Kit for intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.