Please login to your online account to display your discounted pricing

Human/Mouse/Rat Total GSK-3 alpha/beta DuoSet IC, R&D Systems

$428.87 - $428.87

Specifications

Applications Development of sandwich ELISA to measure GSK-3 alpha/beta in cell lysates
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 kit
Species Human, mouse, rat
View More Specs
Products
Catalog Number Mfr. No. Format Price Quantity    

DYC21572

 
r & d systems
RDSDYC21572
Two 96-well plates Each for $428.87
Description & Specifications

Specifications

Applications Development of sandwich ELISA to measure GSK-3 alpha/beta in cell lysates
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 kit
Species Human, mouse, rat

Assay Type: ELISA Development Kit for intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.