Please login to your online account to display your discounted pricing

Human/Mouse/Rat Total GSK-3 alpha/beta DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC21572

 View more versions of this product

Catalog No. DYC21572

  • / Each
 

Description & Specifications

Specifications

Product Type ELISA kit
Applications Development of sandwich ELISA to measure GSK-3 alpha/beta in cell lysates
Quantity 1 kit
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Species Human, mouse, rat
Format Two 96-well plates