Please login to your online account to display your discounted pricing

Human/Mouse/Rat Total GAPDH DuoSet IC, R&D Systems

$428.87 - $846.01

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Quantity 1 Kit
Species Human, Mouse, Rat
View More Specs
Products
Catalog Number Mfr. No. Gene ID (Entrez) Format Price Quantity    

DYC57185

 
r & d systems
RDSDYC57185
24383 Five 96-well plates Each for $846.01

DYC57182

 
r & d systems
RDSDYC57182
24383 Two 96-well plates Each for $428.87
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Quantity 1 Kit
Species Human, Mouse, Rat

Assay Type: ELISA Development Kit for intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.