Please login to your online account to display your discounted pricing

Human/Mouse/Rat Total Akt1 DuoSet IC, R&D Systems

$419.81 - $1,780.05

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 207
Species Human, Mouse, Rat
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC1775E

 
r & d systems
RDSDYC1775E
15 Plates Each for $1,780.05

DYC17752

 
r & d systems
RDSDYC17752
2 Plates Each for $419.81

DYC17755

 
r & d systems
RDSDYC17755
5 Plates Each for $828.14
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 207
Species Human, Mouse, Rat

Assay Type: ELISA Development Kit for intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.