Please login to your online account to display your discounted pricing

Human/Mouse Phospho-EphA5 DuoSet IC, R&D Systems

$419.81 - $419.81

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 13839
Species Human, Mouse
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC50372

 
r & d systems
RDSDYC50372
2 Plates Each for $419.81
Description & Specifications

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 13839
Species Human, Mouse

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins